4-[(2-Fluoro-4-nitrophenyl)(methyl)amino]piperidine, N1-BOC protected
Dodavatel: Apollo Scientific
Synonyms:
tert-Butyl 4-[(2-fluoro-4-nitrophenyl)(methyl)amino]piperidine-1-carboxylate
PC3363-1G
PC3363-250MG
APOSPC3363-1GEA
9530
CZK
APOSPC3363-1G
APOSPC3363-250MG
4-[(2-Fluoro-4-nitrophenyl)(methyl)amino]piperidine, N1-BOC protected
4-[(2-Fluoro-4-nitrophenyl)(methyl)amino]piperidine, N1-BOC protected
Vzorec:
C₁₇H₂₄FN₃O₄ MW: 353.389 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09952120 CAS číslo: 952285-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...