Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoic acid, methyl ester 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
HFPO hexamer methyl ester
PC3336-25G
PC3336-5G
APOSPC3336-25GEA
18020
CZK
APOSPC3336-25G
APOSPC3336-5G
Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoic acid, methyl ester 97%
Perfluoro-2,5,8,11,14-pentamethyl-3,6,9,12,15-pentaoxaoctadecanoic acid, methyl ester
Vzorec:
C₁₉H₃F₃₅O₇ Bod varu: 97…98 °C (0,6 mm Hg) Hustota: 1,8 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD04038871 CAS číslo: 1005033-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...