1-[4-(Difluoromethoxy)phenyl]-imidazolidin-2-one
Dodavatel: Apollo Scientific
|
Varování
|
PC32982-100MG
PC32982-1G
PC32982-250MG
APOSPC32982-100MGEA
622
CZK
APOSPC32982-100MG
APOSPC32982-1G
APOSPC32982-250MG
1-[4-(Difluoromethoxy)phenyl]-imidazolidin-2-one
1-[4-(Difluoromethoxy)phenyl]-imidazolidin-2-one
Vzorec:
C₁₀H₁₀F₂N₂O₂ MW: 228,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24189778 CAS číslo: 1550059-18-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...