Tisk…

1-(3-Chloro-4-fluorophenyl)-imidazolidin-2-one

Dodavatel: Apollo Scientific
Varování

PC32980-1G PC32980-250MG
APOSPC32980-1GEA 2400 CZK
APOSPC32980-1G APOSPC32980-250MG
1-(3-Chloro-4-fluorophenyl)-imidazolidin-2-one
1-(3-Chloro-4-fluorophenyl)-imidazolidin-2-one
Vzorec: C₉H₈ClFN₂O
MW: 214,624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23696674
CAS číslo: 848759-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR