1-(3,4-Difluorophenyl)-imidazolidin-2-one
Dodavatel: Apollo Scientific
|
Varování
|
PC32977-1G
PC32977-250MG
APOSPC32977-1GEA
2400
CZK
APOSPC32977-1G
APOSPC32977-250MG
1-(3,4-Difluorophenyl)-imidazolidin-2-one
1-(3,4-Difluorophenyl)-imidazolidin-2-one
Vzorec:
C₉H₈F₂N₂O MW: 198,169 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23696668 CAS číslo: 162748-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...