Tisk…

1-(3-Fluorophenyl)-imidazolidin-2-one

Dodavatel: Apollo Scientific
Varování

PC32975-1G PC32975-250MG
APOSPC32975-1GEA 10980 CZK
APOSPC32975-1G APOSPC32975-250MG
1-(3-Fluorophenyl)-imidazolidin-2-one
1-(3-Fluorophenyl)-imidazolidin-2-one
Vzorec: C₉H₉FN₂O
MW: 180,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23696666
CAS číslo: 169547-81-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR