Tisk…

4-(Difluoromethyl)-2-(trifluoromethyl)benzonitrile 95%

Dodavatel: Apollo Scientific
Nebezpečí

PC32953-1G PC32953-250MG PC32953-5G
APOSPC32953-1GEA 2150 CZK
APOSPC32953-1G APOSPC32953-250MG APOSPC32953-5G
4-(Difluoromethyl)-2-(trifluoromethyl)benzonitrile 95%
4-(Difluoromethyl)-2-(trifluoromethyl)benzonitrile
Vzorec: C₉H₄F₅N
MW: 221.127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28790571
CAS číslo: 1807192-90-0
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR