3-Bromo-5-fluoro-1-cyclopropylbenzene
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Bromo-3-cyclopropyl-5-fluorobenzene
PC32949-1G
PC32949-250MG
PC32949-5G
APOSPC32949-1GEA
3580
CZK
APOSPC32949-1G
APOSPC32949-250MG
APOSPC32949-5G
3-Bromo-5-fluoro-1-cyclopropylbenzene
3-Bromo-5-fluoro-1-cyclopropylbenzene
Vzorec:
C₉H₈BrF MW: 215,062 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21099988 CAS číslo: 1311265-74-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...