Tisk…

3-Bromo-4-fluoro-1-cyclopropylbenzene

Dodavatel: Apollo Scientific
Varování

PC32948-1G PC32948-250MG
APOSPC32948-1GEA 3270 CZK
APOSPC32948-1G APOSPC32948-250MG
3-Bromo-4-fluoro-1-cyclopropylbenzene
3-Bromo-4-fluoro-1-cyclopropylbenzene
Vzorec: C₉H₈BrF
MW: 215,062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21099995
CAS číslo: 1353854-28-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR