2-Bromo-3-fluoro-1-cyclopropylbenzene
Dodavatel: Apollo Scientific
|
Varování
|
PC32943-1G
PC32943-250MG
APOSPC32943-1GEA
18620
CZK
APOSPC32943-1G
APOSPC32943-250MG
2-Bromo-3-fluoro-1-cyclopropylbenzene
2-Bromo-3-fluoro-1-cyclopropylbenzene
Vzorec:
C₉H₈BrF Storage Temperature: Ambient |
MDL Number:
MFCD24449791 CAS číslo: 1434127-29-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...