2-(Difluoromethyl)-5-chloro-1H-benzimidazole
Dodavatel: Apollo Scientific
|
Varování
|
PC32937-1G
PC32937-250MG
PC32937-5G
APOSPC32937-1GEA
14880
CZK
APOSPC32937-1G
APOSPC32937-250MG
APOSPC32937-5G
2-(Difluoromethyl)-5-chloro-1H-benzimidazole
2-(Difluoromethyl)-5-chloro-1H-benzimidazole
Vzorec:
C₈H₅ClF₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD20390822 CAS číslo: 91437-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...