5-Bromo-2-(difluoromethyl)-1H-benzimidazole
Dodavatel: Apollo Scientific
|
Varování
|
PC32935-1G
PC32935-250MG
APOSPC32935-1GEA
16340
CZK
APOSPC32935-1G
APOSPC32935-250MG
5-Bromo-2-(difluoromethyl)-1H-benzimidazole
5-Bromo-2-(difluoromethyl)-1H-benzimidazole
Vzorec:
C₈H₅BrF₂N₂ Storage Temperature: Ambient |
MDL Number:
MFCD20403496 CAS číslo: 1256956-70-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...