Tisk…

2-[Bromo(difluoro)methyl]-5-nitro-1H-benzimidazole

Dodavatel: Apollo Scientific
Varování

PC32930-1G PC32930-250MG PC32930-5G
APOSPC32930-1GEA 1210 CZK
APOSPC32930-1G APOSPC32930-250MG APOSPC32930-5G
2-[Bromo(difluoro)methyl]-5-nitro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-5-nitro-1H-benzimidazole
Vzorec: C₈H₄BrF₂N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD31922513
CAS číslo: 1353778-45-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR