Tisk…

2-[Bromo(difluoro)methyl]-5-chloro-1H-benzimidazole

Dodavatel: Apollo Scientific
Varování

PC32922-1G PC32922-250MG PC32922-5G
APOSPC32922-1GEA 1660 CZK
APOSPC32922-1G APOSPC32922-250MG APOSPC32922-5G
2-[Bromo(difluoro)methyl]-5-chloro-1H-benzimidazole
2-[Bromo(difluoro)methyl]-5-chloro-1H-benzimidazole
Vzorec: C₈H₄BrClF₂N₂
MW: 281,485 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31922505
CAS číslo: 1353778-40-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR