Tisk…

4-(3-Chloro-4-fluorophenyl)-1H-1,2,4-triazol-5(4H)-one

Dodavatel: Apollo Scientific
Varování

PC32900-1G PC32900-250MG
APOSPC32900-1GEA 2400 CZK
APOSPC32900-1G APOSPC32900-250MG
4-(3-Chloro-4-fluorophenyl)-1H-1,2,4-triazol-5(4H)-one
4-(3-Chloro-4-fluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Vzorec: C₈H₅ClFN₃O
MW: 213,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31742298
CAS číslo: 1346235-03-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR