Tisk…

4-(3,5-Difluorophenyl)-1H-1,2,4-triazol-5(4H)-one

Dodavatel: Apollo Scientific
Varování

PC32897-1G PC32897-250MG
APOSPC32897-1GEA 2400 CZK
APOSPC32897-1G APOSPC32897-250MG
4-(3,5-Difluorophenyl)-1H-1,2,4-triazol-5(4H)-one
4-(3,5-Difluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Vzorec: C₈H₅F₂N₃O
MW: 197,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31742300
CAS číslo: 1817687-97-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR