4-(3,4-Difluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Dodavatel: Apollo Scientific
|
Varování
|
PC32896-1G
PC32896-250MG
APOSPC32896-1GEA
2400
CZK
APOSPC32896-1G
APOSPC32896-250MG
4-(3,4-Difluorophenyl)-1H-1,2,4-triazol-5(4H)-one
4-(3,4-Difluorophenyl)-1H-1,2,4-triazol-5(4H)-one
Vzorec:
C₈H₅F₂N₃O MW: 197,142 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31742299 CAS číslo: 1817687-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...