Tisk…

4-(3-Fluorophenyl)-3-methyl-1H-1,2,4-triazol-5-one

Dodavatel: Apollo Scientific
Varování

PC32894-1G PC32894-250MG
APOSPC32894-1GEA 29760 CZK
APOSPC32894-1G APOSPC32894-250MG
4-(3-Fluorophenyl)-3-methyl-1H-1,2,4-triazol-5-one
4-(3-Fluorophenyl)-3-methyl-1H-1,2,4-triazol-5-one
Vzorec: C₉H₈FN₃O
Storage Temperature: Ambient
MDL Number: MFCD03787480
CAS číslo: 860650-60-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR