Tisk…

6-Fluorobenzofuran

Dodavatel: Apollo Scientific
Varování

PC32891-1G PC32891-250MG
APOSPC32891-1GEA 16370 CZK
APOSPC32891-1G APOSPC32891-250MG
6-Fluorobenzofuran
6-Fluorobenzofuran
Vzorec: C₈H₅FO
MW: 136,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28654686
CAS číslo: 24410-60-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR