Tisk…

(2,2-Difluorocyclopropyl)methanamine hydrochloride

Dodavatel: Apollo Scientific
Varování

PC32886-1G PC32886-250MG PC32886-5G
APOSPC32886-1GEA 17460 CZK
APOSPC32886-1G APOSPC32886-250MG APOSPC32886-5G
(2,2-Difluorocyclopropyl)methanamine hydrochloride
(2,2-Difluorocyclopropyl)methanamine hydrochloride
Vzorec: C₄H₇F₂N·ClH
MW: 143,563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26407844
CAS číslo: 1426309-49-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR