Tisk…

5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl

Dodavatel: Apollo Scientific

PC32869-1G PC32869-250MG PC32869-500MG PC32869-2.5G
APOSPC32869-1GEA 14150 CZK
APOSPC32869-1G APOSPC32869-250MG APOSPC32869-500MG APOSPC32869-2.5G
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl
Vzorec: C₆H₃Cl₂F₃N₂O
MW: 247,002 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115396
CAS číslo: 128564-57-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR