Tisk…

(2-Bromo-4,5-difluorophenyl)methanamine

Dodavatel: Apollo Scientific

PC32856-1G PC32856-250MG PC32856-500MG PC32856-2.5G
APOSPC32856-1GEA 18820 CZK
APOSPC32856-1G APOSPC32856-250MG APOSPC32856-500MG APOSPC32856-2.5G
(2-Bromo-4,5-difluorophenyl)methanamine
(2-Bromo-4,5-difluorophenyl)methanamine
Vzorec: C₇H₆BrF₂N
MW: 222,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546120
CAS číslo: 1256238-50-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR