Tisk…
3-(3-Bromo-4-fluorophenyl)propionic acid 95%
Dodavatel: Apollo Scientific
Synonyms: 3-(3-Bromo-4-fluorophenyl)propanoic acid
PC32847-1G PC32847-5G
APOSPC32847-1GEA 1700 CZK
APOSPC32847-1G APOSPC32847-5G
3-(3-Bromo-4-fluorophenyl)propionic acid 95%
3-(3-Bromo-4-fluorophenyl)propionic acid
Vzorec: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12190913
CAS číslo: 866862-24-0