Tisk…

4'-Bromo-2',6'-difluoroacetophenone 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(4-Bromo-2,6-difluorophenyl)ethanone, 1-(4-Bromo-2,6-difluorophenyl)ethan-1-one

PC32806-250MG PC32806-1G PC32806-5G
APOSPC32806-250MGEA 622 CZK
APOSPC32806-250MG APOSPC32806-1G APOSPC32806-5G
4'-Bromo-2',6'-difluoroacetophenone 97%
4'-Bromo-2',6'-difluoroacetophenone
Vzorec: C₈H₅BrF₂O
MW: 235,026 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22570410
CAS číslo: 746630-34-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR