8-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
PC32790-1G
PC32790-250MG
APOSPC32790-1GEA
4470
CZK
APOSPC32790-1G
APOSPC32790-250MG
8-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
8-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Vzorec:
C₁₀H₅FO₄ MW: 208,143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13187194 CAS číslo: 625823-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...