1-Bromo-2-chloro-3,5-difluorobenzene 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC32780-10G
PC32780-1G
PC32780-5G
APOSPC32780-10GEA
2940
CZK
APOSPC32780-10G
APOSPC32780-1G
APOSPC32780-5G
1-Bromo-2-chloro-3,5-difluorobenzene 98%
1-Bromo-2-chloro-3,5-difluorobenzene
Vzorec:
C₆H₂BrClF₂ MW: 227,434 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00135441 CAS číslo: 187929-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...