Tisk…

5-Fluoro-2-cyclopropylphenol

Dodavatel: Apollo Scientific
Varování

PC32779-1G PC32779-250MG
APOSPC32779-1GEA 1990 CZK
APOSPC32779-1G APOSPC32779-250MG
5-Fluoro-2-cyclopropylphenol
5-Fluoro-2-cyclopropylphenol
Vzorec: C₉H₉FO
Storage Temperature: Freezer
MDL Number: MFCD25994492
CAS číslo: 1557096-27-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR