4-Fluoro-2-cyclopropylphenol
Dodavatel: Apollo Scientific
|
Varování
|
PC32777-1G
PC32777-250MG
APOSPC32777-1GEA
20470
CZK
APOSPC32777-1G
APOSPC32777-250MG
4-Fluoro-2-cyclopropylphenol
4-Fluoro-2-cyclopropylphenol
Vzorec:
C₉H₉FO MW: 152,166 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD24719781 CAS číslo: 1232774-27-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...