3-Fluoro-4-cyclopropylphenol
Dodavatel: Apollo Scientific
|
Varování
|
PC32775-1G
PC32775-250MG
APOSPC32775-1GEA
18620
CZK
APOSPC32775-1G
APOSPC32775-250MG
3-Fluoro-4-cyclopropylphenol
3-Fluoro-4-cyclopropylphenol
Vzorec:
C₉H₉FO MW: 152,166 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD25994495 CAS číslo: 1545520-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...