3-Fluoro-2-cyclopropylphenol
Dodavatel: Apollo Scientific
|
Varování
|
PC32774-1G
PC32774-250MG
APOSPC32774-1GEA
1990
CZK
APOSPC32774-1G
APOSPC32774-250MG
3-Fluoro-2-cyclopropylphenol
3-Fluoro-2-cyclopropylphenol
Vzorec:
C₉H₉FO MW: 152,166 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD31700581 CAS číslo: 2379918-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...