Tisk…

2-Fluoro-4-cyclopropylphenol

Dodavatel: Apollo Scientific
Varování

PC32771-1G PC32771-250MG
APOSPC32771-1GEA 1080 CZK
APOSPC32771-1G APOSPC32771-250MG
2-Fluoro-4-cyclopropylphenol
2-Fluoro-4-cyclopropylphenol
Vzorec: C₉H₉FO
Storage Temperature: Freezer
MDL Number: MFCD25994499
CAS číslo: 1544937-92-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR