2-Fluoro-3-cyclopropylphenol
Dodavatel: Apollo Scientific
|
Varování
|
PC32770-1G
PC32770-250MG
APOSPC32770-1GEA
1990
CZK
APOSPC32770-1G
APOSPC32770-250MG
2-Fluoro-3-cyclopropylphenol
2-Fluoro-3-cyclopropylphenol
Vzorec:
C₉H₉FO Storage Temperature: Freezer |
MDL Number:
MFCD31700580 CAS číslo: 2290421-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...