Tisk…

tert-Butyl (S)-(1-fluoropropan-2-yl)carbamate 95%

Dodavatel: Apollo Scientific
Varování

PC32766-1G PC32766-250MG
APOSPC32766-1GEA 29760 CZK
APOSPC32766-1G APOSPC32766-250MG
tert-Butyl (S)-(1-fluoropropan-2-yl)carbamate 95%
tert-Butyl (S)-(1-fluoropropan-2-yl)carbamate
Vzorec: C₈H₁₆FNO₂
Storage Temperature: Ambient
MDL Number: MFCD30378942
CAS číslo: 1187926-63-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR