2-Fluoro-6-formylbenzeneboronic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Fluoro-6-formylphenyl)boronic acid, 2-Borono-3-fluorobenzaldehyde, 2-Fluoro-6-formylphenylboronic acid
PC32765-1G
PC32765-5G
PC32765-25G
APOSPC32765-1GEA
1910
CZK
APOSPC32765-1G
APOSPC32765-5G
APOSPC32765-25G
2-Fluoro-6-formylbenzeneboronic acid
2-Fluoro-6-formylbenzeneboronic acid
Vzorec:
C₇H₆BFO₃ MW: 167.93 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD19981524 CAS číslo: 1938062-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...