2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)anisole 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC32761-100G
PC32761-25G
PC32761-5G
APOSPC32761-100GEA
21160
CZK
APOSPC32761-100G
APOSPC32761-25G
APOSPC32761-5G
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)anisole 98%
2-(2-Bromo-1,1,2,2-tetrafluoroethoxy)anisole
Vzorec:
C₉H₇BrF₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD28040491 CAS číslo: 220004-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...