Tisk…

4-Fluoro-2-methyl-5-nitrophenyl trifluoromethanesulphonate 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Fluoro-4-nitro-2-{[(trifluoromethyl)sulphonyl]oxy}toluene, 4-Fluoro-2-methyl-5-nitrophenyl triflate

PC32754-1G PC32754-250MG
APOSPC32754-1GEA 5530 CZK
APOSPC32754-1G APOSPC32754-250MG
4-Fluoro-2-methyl-5-nitrophenyl trifluoromethanesulphonate 95%
4-Fluoro-2-methyl-5-nitrophenyl trifluoromethanesulphonate
Vzorec: C₈H₅F₄NO₅S
MW: 303,188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27937148
CAS číslo: 1820734-74-4
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR