Tisk…

2-Nitro-6-(trifluoromethyl)benzonitrile

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Cyano-3-(trifluoromethyl)nitrobenzene, 2-Cyano-3-nitrobenzotrifluoride, 6-Nitro-α,α,α-trifluoro-o-tolunitrile

PC32747-1G PC32747-5G PC32747-25G
APOSPC32747-1GEA 622 CZK
APOSPC32747-1G APOSPC32747-5G APOSPC32747-25G
2-Nitro-6-(trifluoromethyl)benzonitrile
2-Nitro-6-(trifluoromethyl)benzonitrile
Vzorec: C₈H₃F₃N₂O₂
MW: 216,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16606330
CAS číslo: 1227581-80-7
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR