Tisk…

3-Fluoro-4-iodobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: α-Bromo-3-fluoro-4-iodotoluene, 4-(Bromomethyl)-2-fluoro-1-iodobenzene

PC32738-1G PC32738-250MG PC32738-5G
APOSPC32738-1GEA 8530 CZK
APOSPC32738-1G APOSPC32738-250MG APOSPC32738-5G
3-Fluoro-4-iodobenzyl bromide
3-Fluoro-4-iodobenzyl bromide
Vzorec: C₇H₅BrFI
MW: 314,921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396959
CAS číslo: 1022931-83-4
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR