Tisk…

3-Fluoro-2-iodobenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(Bromomethyl)-3-fluoro-2-iodobenzene, α-Bromo-3-fluoro-2-iodotoluene

PC32737-1G PC32737-250MG PC32737-5G
APOSPC32737-1GEA 8200 CZK
APOSPC32737-1G APOSPC32737-250MG APOSPC32737-5G
3-Fluoro-2-iodobenzyl bromide
3-Fluoro-2-iodobenzyl bromide
Vzorec: C₇H₅BrFI
MW: 314,921 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396958
CAS číslo: 1261675-04-0
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR