Tisk…

N-Benzyl-N-[2-(trifluoromethoxy)benzylidene]amine

Dodavatel: Apollo Scientific

PC32476-1G PC32476-250MG
APOSPC32476-1GEA 2820 CZK
APOSPC32476-1G APOSPC32476-250MG
N-Benzyl-N-[2-(trifluoromethoxy)benzylidene]amine
N-Benzyl-N-[2-(trifluoromethoxy)benzylidene]amine
Vzorec: C₁₅H₁₂F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09998151
CAS číslo: 259251-79-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR