N2,N2-Dimethyl-3-(trifluoromethyl)pyridine-2,5-diamine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-Amino-2-(dimethylamino)-α,α,α-trifluoro-3-picoline, 5-Amino-2-(dimethylamino)-3-(trifluoromethyl)pyridine
PC32086-250MG
PC32086-1G
APOSPC32086-250MGEA
14320
CZK
APOSPC32086-250MG
APOSPC32086-1G
N2,N2-Dimethyl-3-(trifluoromethyl)pyridine-2,5-diamine
N2,N2-Dimethyl-3-(trifluoromethyl)pyridine-2,5-diamine
Vzorec:
C₈H₁₀F₃N₃ MW: 205.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02090020 CAS číslo: 288247-16-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...