Tisk…

N-Ethyl-2-fluoroaniline 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(2-Fluorophenyl)ethylamine

PC3207-10G PC3207-1G PC3207-5G
APOSPC3207-10GEA 20400 CZK
APOSPC3207-10G APOSPC3207-1G APOSPC3207-5G
N-Ethyl-2-fluoroaniline 95%
N-Ethyl-2-fluoroaniline
Vzorec: C₈H₁₀FN
MW: 139,17 g/mol
Bod varu: 72…74 °C (15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00203991
CAS číslo: 2707-64-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR