2,3-Difluoro-DL-phenylglycine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Amino-2-(2,3-difluorophenyl)ethanoic acid, Amino(2,3-difluorophenyl)acetic acid
PC3203-1G
PC3203-5G
APOSPC3203-1GEA
2070
CZK
APOSPC3203-1G
APOSPC3203-5G
2,3-Difluoro-DL-phenylglycine
2,3-Difluoro-DL-phenylglycine
Vzorec:
C₈H₇F₂NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD06660156 CAS číslo: 237424-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...