Tisk…

3-Bromo-N-(2-fluorophenyl)propanamide

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-N-(2-fluorophenyl)propionamide, N-(3-Bromopropanoyl)-2-fluoroaniline

PC31961-1G PC31961-250MG
APOSPC31961-1GEA 17200 CZK
APOSPC31961-1G APOSPC31961-250MG
3-Bromo-N-(2-fluorophenyl)propanamide
3-Bromo-N-(2-fluorophenyl)propanamide
Vzorec: C₉H₉BrFNO
MW: 246,076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00124372
CAS číslo: 7661-09-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR