[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-Fluoro-4-[4-(hydroxymethyl)-5-methyl-1H-pyrazol-1-yl]benzene, 1-(4-Fluorophenyl)-4-(hydroxymethyl)-5-methyl-1H-pyrazole
PC31886-1G
PC31886-250MG
APOSPC31886-1GEA
4170
CZK
APOSPC31886-1G
APOSPC31886-250MG
[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol 97%
[1-(4-Fluorophenyl)-5-methyl-1H-pyrazol-4-yl]methanol
Vzorec:
C₁₁H₁₁FN₂O MW: 206,216 g/mol Bod tání: 120…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD03644148 CAS číslo: 465514-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...