8-Bromo-4-chloro-2-(trifluoromethyl)quinoline
Dodavatel: Apollo Scientific
|
Varování
|
PC31854-250MG
PC31854-1G
PC31854-5G
APOSPC31854-250MGEA
787
CZK
APOSPC31854-250MG
APOSPC31854-1G
APOSPC31854-5G
8-Bromo-4-chloro-2-(trifluoromethyl)quinoline
8-Bromo-4-chloro-2-(trifluoromethyl)quinoline
Vzorec:
C₁₀H₄BrClF₃N Storage Temperature: Ambient |
MDL Number:
MFCD00174293 CAS číslo: 655235-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...