Tisk…

2-(Methylthio)-4-(trifluoromethyl)pyrimidine

Dodavatel: Apollo Scientific

PC31716-1G PC31716-5G
APOSPC31716-1GEA 5290 CZK
APOSPC31716-1G APOSPC31716-5G
2-(Methylthio)-4-(trifluoromethyl)pyrimidine
2-(Methylthio)-4-(trifluoromethyl)pyrimidine
Vzorec: C₆H₅F₃N₂S
MW: 194,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209107
CAS číslo: 136547-20-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR