3-(2-Fluorobenzoyl)-1H-pyrrole
Dodavatel: Apollo Scientific
Synonyms:
(2-Fluorophenyl)-(1H-pyrrol-3-yl)methanone
PC3152-1G
PC3152-500MG
APOSPC3152-1GEA
6720
CZK
APOSPC3152-1G
APOSPC3152-500MG
3-(2-Fluorobenzoyl)-1H-pyrrole
3-(2-Fluorobenzoyl)-1H-pyrrole
Vzorec:
C₁₁H₈FNO MW: 189,186 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08056192 CAS číslo: 198126-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...