2-(2-Chloro-6-fluorobenzylthio)aniline
Dodavatel: Apollo Scientific
|
Varování
|
PC31505-1G
PC31505-250MG
PC31505-5G
APOSPC31505-1GEA
4570
CZK
APOSPC31505-1G
APOSPC31505-250MG
APOSPC31505-5G
2-(2-Chloro-6-fluorobenzylthio)aniline
2-(2-Chloro-6-fluorobenzylthio)aniline
Vzorec:
C₁₃H₁₁ClFNS MW: 267,75 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052213 CAS číslo: 646989-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...