Ethyl 2-{[(trifluoromethyl)sulphonyl]oxy}cyclopent-1-ene-1-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Ethoxycarbonyl)cyclopent-1-en-1-yl triflate, 2-(Ethoxycarbonyl)cyclopent-1-en-1-yl trifluoromethanesulphonate
PC31418-1G
PC31418-250MG
PC31418-5G
APOSPC31418-1GEA
5390
CZK
APOSPC31418-1G
APOSPC31418-250MG
APOSPC31418-5G
Ethyl 2-{[(trifluoromethyl)sulphonyl]oxy}cyclopent-1-ene-1-carboxylate
Ethyl 2-{[(trifluoromethyl)sulphonyl]oxy}cyclopent-1-ene-1-carboxylate
Vzorec:
C₉H₁₁F₃O₅S MW: 288,241 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00209586 CAS číslo: 122539-74-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...